MilliporeSigma
Signaling of type II oncostatin M receptor.
The Journal of biological chemistry (1997-06-20)
P Auguste, C Guillet, M Fourcin, C Olivier, J Veziers, A Pouplard-Barthelaix, H Gascan
ABSTRACT
Oncostatin M (OSM) mediates its bioactivities through two different heterodimer receptors. They both involve the gp130-transducing receptor, which dimerizes with either leukemia inhibitory receptor beta or with OSM receptor beta (OSMRbeta) to generate, respectively, type I and type II OSM receptors. Co-precipitation of gp130-associated proteins, flow cytometry, polymerase chain reaction, and tyrosine phosphorylation analyses allowed the characterization of both types of OSM receptors expressed on the surface of different cell lines. It also allowed the detection of a large size protein, p250, that specifically associates to the type II OSM receptor components and that is tyrosine-phosphorylated after the activation peak of the gp130.OSMRbeta heterocomplex. The restricted expression of type I OSM receptor by the JAR choriocarcinoma cell line, and type II receptor by the A375 melanoma cell line, permitted the characterization of their signaling machineries. Both type I and type II OSM receptors activated Jak1, Jak2, and Tyk2 receptor-associated tyrosine kinases. The information is next relayed to the nucleus by the STAT3 transcriptional activator, which is recruited by both types of OSM receptors. In addition, STAT5b was specifically activated through the gp130.OSMRbeta type II heterocomplex. The signaling pathway differences observed between the common type I LIF/OSM receptor and the specific type II OSM receptor might explain some of the bioactivities specifically displayed by OSM.