MilliporeSigma
  • cDNA cloning and sequence analysis of the bovine adrenocorticotropic hormone (ACTH) receptor.
cDNA cloning and sequence analysis of the bovine adrenocorticotropic hormone (ACTH) receptor.
Biochimica et biophysica acta (1994-02-17)
M Raikhinstein, M Zohar, I Hanukoglu
ABSTRACT
We isolated five independent cDNAs of nearly 3000 bp for the bovine ACTH receptor by screening adrenal cortex cDNA libraries with a PCR cloned cDNA fragment. The deduced receptor sequence includes 297 residues (M(r) = 33,258) with 81% identity with the human ACTH receptor, and shows seven hydrophobic transmembrane domains. The calculated M(r) of the receptor is smaller than the 40-45 kDa observed in crosslinking studies with labeled ACTH. Since the bovine and human receptors have two glycosylation motifs in the N-terminus, the difference may result from glycosylation of the receptor. Analysis of the sequences of both bovine and human receptors revealed a single protein kinase. A phosphorylation motif located in the third intracellular loop (Ser-209) juxtaposed to a protein kinase C phosphorylation motif (Thr-204). Thus, the involvement of protein kinase A and C pathways in ACTH action may be mediated in part by phosphorylation of the ACTH receptor at these motifs. The 3'-untranslated region of the bovine cDNA is > 2000 bp and includes two inverse repeats giving an extensive and strong secondary structure to the ACTH receptor RNA.