MilliporeSigma
  • Direct conversion of fibroblasts to neurons by reprogramming PTB-regulated microRNA circuits.
Direct conversion of fibroblasts to neurons by reprogramming PTB-regulated microRNA circuits.
Cell (2013-01-15)
Yuanchao Xue, Kunfu Ouyang, Jie Huang, Yu Zhou, Hong Ouyang, Hairi Li, Gang Wang, Qijia Wu, Chaoliang Wei, Yanzhen Bi, Li Jiang, Zhiqiang Cai, Hui Sun, Kang Zhang, Yi Zhang, Ju Chen, Xiang-Dong Fu
ABSTRACT
The induction of pluripotency or trans-differentiation of one cell type to another can be accomplished with cell-lineage-specific transcription factors. Here, we report that repression of a single RNA binding polypyrimidine-tract-binding (PTB) protein, which occurs during normal brain development via the action of miR-124, is sufficient to induce trans-differentiation of fibroblasts into functional neurons. Besides its traditional role in regulated splicing, we show that PTB has a previously undocumented function in the regulation of microRNA functions, suppressing or enhancing microRNA targeting by competitive binding on target mRNA or altering local RNA secondary structure. A key event during neuronal induction is the relief of PTB-mediated blockage of microRNA action on multiple components of the REST complex, thereby derepressing a large array of neuronal genes, including miR-124 and multiple neuronal-specific transcription factors, in nonneuronal cells. This converts a negative feedback loop to a positive one to elicit cellular reprogramming to the neuronal lineage.