MilliporeSigma
  • Tumor-infiltrating DCs suppress nucleic acid-mediated innate immune responses through interactions between the receptor TIM-3 and the alarmin HMGB1.
Tumor-infiltrating DCs suppress nucleic acid-mediated innate immune responses through interactions between the receptor TIM-3 and the alarmin HMGB1.
Nature immunology (2012-07-31)
Shigeki Chiba, Muhammad Baghdadi, Hisaya Akiba, Hironori Yoshiyama, Ichiro Kinoshita, Hirotoshi Dosaka-Akita, Yoichiro Fujioka, Yusuke Ohba, Jacob V Gorman, John D Colgan, Mitsuomi Hirashima, Toshimitsu Uede, Akinori Takaoka, Hideo Yagita, Masahisa Jinushi
ABSTRACT
The mechanisms by which tumor microenvironments modulate nucleic acid-mediated innate immunity remain unknown. Here we identify the receptor TIM-3 as key in circumventing the stimulatory effects of nucleic acids in tumor immunity. Tumor-associated dendritic cells (DCs) in mouse tumors and patients with cancer had high expression of TIM-3. DC-derived TIM-3 suppressed innate immune responses through the recognition of nucleic acids by Toll-like receptors and cytosolic sensors via a galectin-9-independent mechanism. In contrast, TIM-3 interacted with the alarmin HMGB1 to interfere with the recruitment of nucleic acids into DC endosomes and attenuated the therapeutic efficacy of DNA vaccination and chemotherapy by diminishing the immunogenicity of nucleic acids released from dying tumor cells. Our findings define a mechanism whereby tumor microenvironments suppress antitumor immunity mediated by nucleic acids.