MilliporeSigma
  • WAC, a functional partner of RNF20/40, regulates histone H2B ubiquitination and gene transcription.
WAC, a functional partner of RNF20/40, regulates histone H2B ubiquitination and gene transcription.
Molecular cell (2011-02-19)
Feng Zhang, Xiaochun Yu
ABSTRACT
Histone H2B ubiquitination plays an important role in regulating chromatin organization during gene transcription. It has been shown that RNF20/40 regulates H2B ubiquitination. Here, using protein affinity purification, we have identified WAC as a functional partner of RNF20/40. Depletion of WAC abolishes H2B ubiquitination. WAC interacts with RNF20/40 through its C-terminal coiled-coil region and promotes RNF20/40 s E3 ligase activity for H2B ubiquitination. The N-terminal WW domain of WAC recognizes RNA polymerase II. During gene transcription, WAC targets RNF20/40 to associate with RNA polymerase II complex for H2B ubiquitination at active transcription sites, which regulates transcription. Moreover, WAC-dependent transcription is important for cell-cycle checkpoint activation in response to genotoxic stress. Taken together, our results demonstrate an important regulator for transcription-coupled histone H2B ubiquitination.