MilliporeSigma
  • CD28-dependent HIV-1 transcription is associated with Vav, Rac, and NF-kappa B activation.
CD28-dependent HIV-1 transcription is associated with Vav, Rac, and NF-kappa B activation.
The Journal of biological chemistry (2003-07-05)
Julie A Cook, Lee Albacker, Avery August, Andrew J Henderson
ABSTRACT
Activation of HIV-1-infected T cells through the T cell receptor and costimulatory molecule CD28 induces proviral transcription; however, the mechanism behind this enhanced virus expression is unknown. Jurkat T cells and primary CD4+ T cells expressing a CD8 alpha/CD28 chimeric receptor containing a mutation at tyrosine 200 in the cytoplasmic tail were unable to fully induce HIV-1 proviral transcription in response to CD8 alpha/28 receptor cross-linking in comparison to CD28 costimulation. The loss of transactivation seen with the mutant chimeric receptor correlated with a decrease in Vav tyrosine phosphorylation. CD28-dependent activation of HIV-1 transcription also required the GTPase activity of Rac1, which was not activated during costimulation with the mutated receptor. Furthermore, the mutated receptor was unable to induce NF-kappa B DNA binding or transactivation, as demonstrated by electromobility shift assays and HIV-1 long terminal repeat and NF-kappa B-dependent reporter constructs. These studies show that signaling events initiated by tyrosine 200 of CD28 are required for efficient expression of HIV-1 transcription in activated T cells.
Sigma-Aldrich
Rac1 Activation Assay Kit, Non-radioactive Rac1 Activation Assay Kit can be used to precipitate Rac1-GTP from cell lysates & detection by a Rac1 specific monoclonal antibody.