MilliporeSigma
  • Microtubule-associated protein MAP1A, MAP1B, and MAP2 proteolysis during soluble amyloid beta-peptide-induced neuronal apoptosis. Synergistic involvement of calpain and caspase-3.
Microtubule-associated protein MAP1A, MAP1B, and MAP2 proteolysis during soluble amyloid beta-peptide-induced neuronal apoptosis. Synergistic involvement of calpain and caspase-3.
The Journal of biological chemistry (2005-10-20)
Alexandre Fifre, Isabelle Sponne, Violette Koziel, Badreddine Kriem, Frances T Yen Potin, Bernard E Bihain, Jean-Luc Olivier, Thierry Oster, Thierry Pillot
ABSTRACT
A growing body of evidence supports the notion that soluble oligomeric forms of the amyloid beta-peptide (Abeta) may be the proximate effectors of neuronal injuries and death in the early stages of Alzheimer disease. However, the molecular mechanisms associated with neuronal apoptosis induced by soluble Abeta remain to be elucidated. We recently demonstrated the involvement of an early reactive oxygen species-dependent perturbation of the microtubule network (Sponne, I., Fifre, A., Drouet, B., Klein, C., Koziel, V., Pincon-Raymond, M., Olivier, J.-L., Chambaz, J., and Pillot, T. (2003) J. Biol. Chem. 278, 3437-3445). Because microtubule-associated proteins (MAPs) are responsible for the polymerization, stabilization, and dynamics of the microtubule network, we investigated whether MAPs might represent the intracellular targets that would enable us to explain the microtubule perturbation involved in soluble Abeta-mediated neuronal apoptosis. The data presented here show that soluble Abeta oligomers induce a time-dependent degradation of MAP1A, MAP1B, and MAP2 involving a perturbation of Ca2+ homeostasis with subsequent calpain activation that, on its own, is sufficient to induce the proteolysis of isoforms MAP2a, MAP2b, and MAP2c. In contrast, MAP1A and MAP1B sequential proteolysis results from the Abeta-mediated activation of caspase-3 and calpain. The prevention of MAP1A, MAP1B, and MAP2 proteolysis by antioxidants highlights the early reactive oxygen species generation in the perturbation of the microtubule network induced by soluble Abeta. These data clearly demonstrate the impact of cytoskeletal perturbations on soluble Abeta-mediated cell death and support the notion of microtubule-stabilizing agents as effective Alzheimer disease drugs.