MilliporeSigma
  • The transcription factor Pax6 regulates survival of dopaminergic olfactory bulb neurons via crystallin αA.
The transcription factor Pax6 regulates survival of dopaminergic olfactory bulb neurons via crystallin αA.
Neuron (2010-11-26)
Jovica Ninkovic, Luisa Pinto, Stefania Petricca, Alexandra Lepier, Jian Sun, Michael A Rieger, Timm Schroeder, Ales Cvekl, Jack Favor, Magdalena Götz
ABSTRACT
Most neurons in the adult mammalian brain survive for the entire life of an individual. However, it is not known which transcriptional pathways regulate this survival in a healthy brain. Here, we identify a pathway regulating neuronal survival in a highly subtype-specific manner. We show that the transcription factor Pax6 expressed in dopaminergic neurons of the olfactory bulb regulates the survival of these neurons by directly controlling the expression of crystallin αA (CryαA), which blocks apoptosis by inhibition of procaspase-3 activation. Re-expression of CryαA fully rescues survival of Pax6-deficient dopaminergic interneurons in vivo and knockdown of CryαA by shRNA in wild-type mice reduces the number of dopaminergic OB interneurons. Strikingly, Pax6 utilizes different DNA-binding domains for its well-known role in fate specification and this role of regulating the survival of specific neuronal subtypes in the mature, healthy brain.