MilliporeSigma
  • Modulation of interleukin (IL)-13 binding and signaling by the gammac chain of the IL-2 receptor.
Modulation of interleukin (IL)-13 binding and signaling by the gammac chain of the IL-2 receptor.
The Journal of biological chemistry (1997-08-08)
N I Obiri, T Murata, W Debinski, R K Puri
ABSTRACT
Interleukin (IL)-13 and IL-4 are cytokine products of TH2 cells which exert similar effects in a variety of cell types. We recently described IL-13R expression on human renal cell and colon carcinoma cells and demonstrated that gammac is not a component of IL-13R or IL-4R systems in these cells. In lymphoid cells such as B cells and monocytes, which respond to IL-13, gammac is a component of IL-4R but does not appear to be a component of IL-13R. Furthermore, while significant IL-13 binding is observed on carcinoma cells, IL-13 barely binds these lymphoid cells and the binding characteristics are different. To better understand the role of gammac in IL-13 binding and signaling, we have transfected a renal cell carcinoma cell line with gammac and examined IL-13 and IL-4 binding and signaling. IL-13 binding as well as IL-13 and IL-4 signaling through the JAK-STAT signaling pathway were severely inhibited. This inhibition was paralleled by a loss of expression of one of the IL-13R chains and intercellular cell adhesion molecule-1. Thus, although gammac has been shown to enhance IL-4 binding and function in some cell types, its influence on IL-13R function in tumor cells appear to be largely negative.