MilliporeSigma
  • AMP-activated protein kinase response to contractions and treatment with the AMPK activator AICAR in young adult and old skeletal muscle.
AMP-activated protein kinase response to contractions and treatment with the AMPK activator AICAR in young adult and old skeletal muscle.
The Journal of physiology (2009-03-11)
D M Thomson, J D Brown, N Fillmore, S K Ellsworth, D L Jacobs, W W Winder, C A Fick, S E Gordon
ABSTRACT
One characteristic of ageing skeletal muscle is a decline in mitochondrial function. Activation of AMP-activated protein kinase (AMPK) occurs in response to an increased AMP/ATP ratio, which is one potential result of mitochondrial dysfunction. We have previously observed higher AMPK activity in old (O; 30 months) vs young adult (YA; 8 months) fast-twitch muscle in response to chronic overload. Here we tested the hypothesis that AMPK would also be hyperactivated in O vs YA fast-twitch extensor digitorum longus muscles from Fischer(344) x Brown Norway (FBN) rats (n = 8 per group) in response to high-frequency electrical stimulation of the sciatic nerve (HFES) or injection of AICAR, an activator of AMPK. Muscles were harvested immediately after HFES (10 sets of six 3-s contractions, 10 s rest between contractions, 1 min rest between sets) or 1 h after AICAR injection (1 mg (g body weight)(-1) subcutaneously). The phosphorylations of AMPKalpha and acetyl-CoA carboxylase (ACC2; a downstream AMPK target) were both greatly increased (P <or= 0.05) in response to HFES in O muscles, but were either unresponsive (AMPK alpha) or much less responsive (ACC) in YA muscles. AMPK alpha2 activity was also greatly elevated in response to HFES in O muscles (but not YA muscles) despite a lower total AMPK alpha2 protein content in O vs YA muscles. In contrast, AMPK alpha2 activity was equally responsive to AICAR treatment in both age groups. Since mitochondrial content and/or efficiency could potentially underlie AMPK hyperactivation, we measured levels of mitochondrial proteins as well as citrate synthase (CS) activity. While CS activity was increased by 25% in O vs YA muscles, uncoupling protein-3 (UCP-3) protein level was upregulated with age by 353%. Thus, AMPK hyperactivation in response to contractile activity in aged fast-twitch muscle may be the result of compromised cellular energetics and not necessarily due to an inherent defect in responsiveness of the AMPK molecule per se.