MilliporeSigma
  • Control of beta1 integrin function. Localization of stimulatory epitopes.
Control of beta1 integrin function. Localization of stimulatory epitopes.
The Journal of biological chemistry (1996-02-09)
J A Wilkins, A Li, H Ni, D G Stupack, C Shen
ABSTRACT
The beta1 integrins can be expressed on the surface of cells in a latent form, which is activated by a variety of stimuli. As an approach to examining the transition to an active receptor, a panel of stimulatory antibodies to beta1 were produced and characterized. These antibodies induced adherence of the T-leukemic cell line Jurkat to collagen and fibronectin. Competitive antibody binding assays indicated the existence of at least three distinct epitope clusters A (B3B11, JB1B, 21C8), B (B44, 13B9), and C(N29) defined by the indicated antibodies. Two antibodies to the A site, JB1B and B3B11, were shown to localize to positions 671-703 and 657-670, respectively, of the beta1. This region is located in an area encompassing a predicted disulfide bond between linearly distant cysteines in beta1 (Cys415-Cys671). The homologous region of the beta3 integrin (490 690 and 602 690) has been shown to be one of the sites recognized by stimulatory antibodies to ligand-induced binding sites. The present results indicate the existence of multiple stimulatory regions and suggest considerable homology between the locations of beta1 and beta3 regulatory sites.