MilliporeSigma
  • Cytokines and hs-CRP levels in individuals treated with low-dose aspirin for cardiovascular prevention: a population-based study (CoLaus Study).
Cytokines and hs-CRP levels in individuals treated with low-dose aspirin for cardiovascular prevention: a population-based study (CoLaus Study).
Cytokine (2014-03-07)
Julien Vaucher, Pedro Marques-Vidal, Gérard Waeber, Peter Vollenweider
ABSTRACT
Pro-inflammatory cytokines and high-sensitive C-reactive protein (hs-CRP) are associated with increased risk for cardiovascular disease. Low-dose aspirin for CV prevention is reported to have anti-inflammatory effects. The aim of this study was to determine the association between pro-inflammatory cytokines and hs-CRP levels and low-dose aspirin use for cardiovascular prevention in a population-based cohort (CoLaus Study). We assessed blood samples in 6085 participants (3201 women) aged 35-75years. Medications' use and indications were recorded. Among aspirin users (n=1'034; 17%), overall low-dose users (351; 5.8%) and low-dose for cardiovascular prevention users (324; 5.3%) were selected for analysis. Pro-inflammatory cytokines (IL-1β, IL-6 and TNF-α were assessed by a multiplex particle-based flow cytometric assay and hs-CRP by an immunometric assay. Cytokines and hs-CRP were presented in quartiles. Multivariate analysis adjusting for sex, age, smoking status, body mass index, diabetes mellitus and immunomodulatory drugs showed no association between cytokines and hs-CRP levels and low-dose aspirin use for cardiovascular prevention, either comparing the topmost vs. the three other quartiles (OR 95% CI, 0.84 (0.59-1.18), 1.03 (0.78-1.32), 1.10 (0.83-1.46), 1.00 (0.67-1.69) for IL-1β, IL-6, TNF-α and hs-CRP, respectively), or comparing the topmost quartile vs. the first one (OR 95% CI, 0.87 (0.60-1.26), 1.19 (0.79-1.79), 1.26 (0.86-1.84), 1.06 (0.67-1.69)). Low-dose aspirin use for cardiovascular prevention does not impact plasma pro-inflammatory cytokine and hs-CRP levels in a population-based cohort.