MilliporeSigma
  • The impact of maternal separation on the number of tyrosine hydroxylase-expressing midbrain neurons during different stages of ontogenesis.
The impact of maternal separation on the number of tyrosine hydroxylase-expressing midbrain neurons during different stages of ontogenesis.
Neuroscience (2011-03-15)
A Chocyk, A Przyborowska, D Dudys, I Majcher, M Maćkowiak, K Wędzony
ABSTRACT
Early life stressors have life-long functional and anatomical consequences. Though many neurotransmitters are involved in the functional impact of early life stress, dopamine seems to be important because of its roles in motor control, adaptation to stressful conditions, mood, cognition, attention and reward. Thus, in the present study, we investigated the way that early life stress, in the form of maternal separation (MS), affects the populations of tyrosine hydroxylase-immunoreactive (TH-IR) dopaminergic neurons in rat midbrain structures during ontogenesis. We included in the study the sub-regions of the substantia nigra (SN) and the ventral tegmental area (VTA). In both the control and MS rats, we found that the estimated total number of TH-expressing neurons fluctuated during ontogenesis. Moreover, MS influenced the number of TH-IR cells, especially in the SN pars reticulata (SNr) and VTA. Shortly after the termination of MS, on postnatal day (PND) 15, a decrease in the estimated total number of TH-IR neurons was observed in the SNr and VTA (in both males and females). On PND 35, MS caused a transient increase in the number of TH-IR cells only in the SNr of female rats. On PND 70, MS affected the number of TH-IR neurons in the VTA of females; specifically, an increase in the number of these cells was observed. Additionally, MS did not alter TH-IR cell sizes or the total levels of TH (measured by Western blot analysis) in the SN and VTA for all stages of ontogenesis in both males and females. The results from the study herein indicate that early life stress has enduring effects on the populations of midbrain TH-expressing dopaminergic neurons (especially in female rats), which are critically important for dopamine-regulated brain function throughout ontogenesis.