MilliporeSigma
  • Extracellular carbohydrate-signal triggering cAMP-dependent protein kinase-dependent neuronal actin-reorganization.
Extracellular carbohydrate-signal triggering cAMP-dependent protein kinase-dependent neuronal actin-reorganization.
Neuroscience (2003-12-04)
N Chen, S Furuya, Y Shinoda, M Yumoto, A Ohtake, K Sato, H Doi, Y Hashimoto, Y Kudo, H Higashi
ABSTRACT
Cell surface glycoconjugates are thought to mediate cell-cell recognition and to play roles in neuronal development and functions. We demonstrated here that exposure of neuronal cells to nanomolar levels of glyco-chains with an N-acetylgalactosamine (GalNAc) residue at the non-reducing termini (GalNAc-S) such as GalNAcbeta4(Neu5Acalpha3)Galbeta4GlcCer (GM2) ganglioside, its oligosaccharide portion, GalNAcbeta4Galbeta4GlcCer (Gg(3)) Cer, GalNAcalpha3GalNAcbeta3Galalpha4Galbeta4GlcCer (Gb(5)) Cer (Forssman hapten) and alpha1-4 linked oligomers of GalNAc, induced a rapid and transient activation of cAMP-dependent protein kinase (PKA) in subplasmalemma. The treatment was accompanied by peripheral actin polymerization and filopodia formation in NG108-15 cells and primary cultured hippocampal neurons, but not in glial cells. A cAMP-dependent protein kinase (PKA) selective inhibitor and an adenylate cyclase inhibitor blocked both PKA activation and the subsequent filopodia formation. A small GTPase cdc42 was a potential downstream target of GalNAc-S-activated PKA. These results suggest that extracellular GalNAc-S serve as potential regulators of the filopodia formation in neuronal cells by triggering the activation of PKA followed by cdc42 up-regulation via a cell surface receptor-like component. Filopodia formation induced by GalNAc-S may have a physiological relevance because long-term exposure to GalNAc-S enhanced F-actin-rich dendrite generation of primary cultured hippocampal neurons, and PKA-dependent dendritic outgrowth and branch formation of primary cultured cerebellar Purkinje neurons, in which actin isoforms were localized to motile structures in dendrites. These findings provide evidence for a novel GalNAc/PKA-signaling cascade in regulating some neuronal maturation.