MilliporeSigma
  • Cutting edge: a novel function for the SLAP-130/FYB adapter protein in beta 1 integrin signaling and T lymphocyte migration.
Cutting edge: a novel function for the SLAP-130/FYB adapter protein in beta 1 integrin signaling and T lymphocyte migration.
Journal of immunology (Baltimore, Md. : 1950) (2000-01-21)
A J Hunter, N Ottoson, N Boerth, G A Koretzky, Y Shimizu
ABSTRACT
The role of integrin-mediated signaling events in T cell function remains incompletely characterized. We report here that alpha4beta1 integrin stimulation of H9 T cells and normal human T cell blasts results in rapid and transient tyrosine phosphorylation of the adapter protein, SH2 domain-containing 76-kDa protein (SLP-76)-associated phosphoprotein of 130 kDa (SLAP-130)/FYB at levels comparable to those observed following TCR stimulation. Stimulation of T cells via the alpha4beta1 integrin enhances the association of tyrosine phosphorylated SLAP-130/FYB with the SH2 domain of the src tyrosine kinase p59fyn. Activation of normal T cells, but not H9 T cells, via alpha4beta1 leads to tyrosine phosphorylation of SLP-76 as well as SLAP-130/FYB. Overexpression of SLAP-130/FYB in normal T cells enhances T cell migration through fibronectin-coated filters in response to the chemokine stromal cell-derived factor (SDF)-1alpha. These results identify SLAP-130/FYB as a new tyrosine phosphorylated substrate in beta1 integrin signaling and suggest a novel function for SLAP-130/FYB in regulating T lymphocyte motility.