MilliporeSigma
  • Conformational and biochemical differences in the TCR.CD3 complex of CD8(+) versus CD4(+) mature lymphocytes revealed in the absence of CD3gamma.
Conformational and biochemical differences in the TCR.CD3 complex of CD8(+) versus CD4(+) mature lymphocytes revealed in the absence of CD3gamma.
The Journal of biological chemistry (1999-11-27)
D A Zapata, A Pacheco-Castro, P S Torres, A R Ramiro, E San José, B Alarcón, L Alibaud, B Rubin, M L Toribio, J R Regueiro
ABSTRACT
Mature CD4(+) and CD8(+) T lymphocytes are believed to build and express essentially identical surface alphabeta T-cell receptor-CD3 (TCR.CD3) complexes. However, TCR.CD3 expression has been shown to be more impaired in CD8(+) cells than in CD4(+) cells when CD3gamma is absent in humans or mice. We have addressed this paradox by performing a detailed phenotypical and biochemical analysis of the TCR.CD3 complex in human CD3gamma-deficient CD8(+) and CD4(+) T cells. The results indicated that the membrane TCR.CD3 complex of CD8(+) T lymphocytes was conformationally different from that of CD4(+) lymphocytes in the absence of CD3gamma. In addition, CD8(+), but not CD4(+), CD3gamma-deficient T lymphocytes were shown to contain abnormally glycosylated TCRbeta proteins, together with a smaller, abnormal TCR chain (probably incompletely processed TCRalpha). These results suggest the existence of hitherto unrecognized biochemical differences between mature CD4(+) and CD8(+) T lymphocytes in the intracellular control of alphabetaTCR. CD3 assembly, maturation, or transport that are revealed when CD3gamma is absent. Such lineage-specific differences may be important in receptor-coreceptor interactions during antigen recognition.