MilliporeSigma
  • Effects of an acidic fibroblast growth factor fragment analog on learning and memory and on medial septum cholinergic neurons in senescence-accelerated mice.
Effects of an acidic fibroblast growth factor fragment analog on learning and memory and on medial septum cholinergic neurons in senescence-accelerated mice.
Neuroscience (1999-07-30)
K Sasaki, I Tooyama, A J Li, Y Oomura, H Kimura
ABSTRACT
We examined the effects of repeated subcutaneous injections of an acidic fibroblast growth factor fragment analog, [Ala16] acidic fibroblast growth factor (1-29), on learning and memory and on the choline acetyltransferase immunoreactivity of forebrain neurons in senescence-accelerated mice. One group of accelerated senescence-prone mice (accelerated senescence-prone-8) received [Ala16] acidic fibroblast growth factor (1-29), whereas the other group of accelerated senescence-prone-8 mice and a group of accelerated senescence-resistant mice (control) received vehicle solution. Injections began at three weeks after birth and were given weekly for 10 months. In a passive avoidance test, the mean retention latency at three, six and nine months of age was significantly longer in controls (vehicle-treated accelerated senescence-resistant-1) and acidic fibroblast growth factor fragment-treated accelerated senescence-prone-8 than in vehicle-treated accelerated senescence-prone-8 mice, and the latency in acidic fibroblast growth factor fragment-treated accelerated senescence-prone-8 mice was significantly shorter than that in controls only at nine months of age. In the Morris water maze task, the mean latency to climb onto the platform was significantly longer in acidic fibroblast growth factor fragment- and vehicle-treated accelerated senescence-prone-8 mice than in controls. However, the mean latency in the third and fourth trial blocks was significantly shorter for acidic fibroblast growth factor fragment-treated accelerated senescence-prone-8 than for vehicle-treated accelerated senescence-prone-8 mice. In the probe trials, controls and acidic fibroblast growth factor fragment-treated accelerated senescence-prone-8 mice spent significantly more time in the quadrant in which the platform had previously been located than in the other three quadrants. In acidic fibroblast growth factor fragment-treated accelerated senescence-prone-8 mice, the density of medial septum neurons intensely stained for choline acetyltransferase was significantly greater than that in vehicle-treated accelerated senescence-prone-8 mice, but significantly less than that in controls. The results indicate that the beneficial effect of [Ala16] acidic fibroblast growth factor (1-29) on learning and memory function in accelerated senescence-prone-8 mice may be related to a preservation of function in medial septum cholinergic neurons.