MilliporeSigma
  • Mutations in the heparin binding domain of fibronectin in cooperation with the V region induce decreases in pp125(FAK) levels plus proteoglycan-mediated apoptosis via caspases.
Mutations in the heparin binding domain of fibronectin in cooperation with the V region induce decreases in pp125(FAK) levels plus proteoglycan-mediated apoptosis via caspases.
The Journal of biological chemistry (1999-10-16)
Y L Kapila, S Wang, P W Johnson
ABSTRACT
Intact fibronectin (FN) protects cells from apoptosis. When FN is fragmented, specific domains induce proteinase expression in fibroblasts. However, it is not known whether specific domains of FN can also regulate apoptosis. We exposed fibroblasts to four recombinant FN fragments and then assayed for apoptosis using criteria of cellular shape change, condensed nuclear morphology, and DNA fragmentation. The fragments extended from the RGD-containing repeat III10 to III15; they included (V(+)) or excluded (V(-)) the alternatively spliced V region and contained either a mutated (H(-)) or an unmutated (H(+)) heparin binding domain. Only the V(+)H(-) fragment triggered decreases in pp125(FAK) levels and apoptosis, which was rescued by intact FN and inhibitors of caspase-1 and caspase-3. This apoptotic mechanism was mediated by a chondroitin sulfate proteoglycan, since treating cells with chondroitin sulfate or chondroitinase reversed the apoptotic cell shape changes. The alpha4 integrin receptor may also be involved, since using a blocking antibody to alpha4 alone induced apoptotic cell shape changes, whereas co-treatment with this antibody plus V(+)H(+) reversed these effects. These results demonstrate that the V and heparin binding domains of FN modulate pp125(FAK) levels and regulate apoptosis through a chondroitin sulfate proteoglycan- and possibly alpha4 integrin-mediated pathway, which triggers a caspase cascade.