MilliporeSigma
  • Transforming growth factor beta induces caspase 3-independent cleavage of alphaII-spectrin (alpha-fodrin) coincident with apoptosis.
Transforming growth factor beta induces caspase 3-independent cleavage of alphaII-spectrin (alpha-fodrin) coincident with apoptosis.
The Journal of biological chemistry (1999-08-07)
T L Brown, S Patil, C D Cianci, J S Morrow, P H Howe
ABSTRACT
Transforming growth factor beta (TGF-beta) is a potent growth inhibitor and inducer of cell death in B-lymphocytes and is essential for immune regulation and maintenance of self-tolerance. In this report the mouse immature B cell line, WEHI 231, was used to examine the mechanisms involved in TGF-beta-mediated apoptosis. Induction of apoptosis is detected as early as 8 h after TGF-beta administration. Coincident with the onset of apoptosis, the cytoskeletal actin-binding protein, alphaII-spectrin (alpha-fodrin) is cleaved into 150-, 115-, and 110-kDa fragments. The broad spectrum caspase inhibitor (Boc-D-fmk (BD-fmk)) completely abolished TGF-beta-induced apoptosis and alphaII-spectrin cleavage. Caspase 3, although present in WEH1 231 cells, was not activated by TGF-beta, nor was its substrate, poly(ADP-ribose) polymerase. These results identify alphaII-spectrin as a novel substrate that is cleaved during TGF-beta-induced apoptosis. Our data provide the first evidence of calpain and caspase 3-independent cleavage of alphaII-spectrin during apoptosis and suggests that TGF-beta induces apoptosis and alphaII-spectrin cleavage via a potentially novel caspase. This report also provides the first direct evidence of caspase 3 activation in WEH1 231 cells and indicates that at least two distinct apoptotic pathways exist.