MilliporeSigma
Heterogeneous intracellular localization and expression of ataxin-3.
Neurobiology of disease (1999-03-09)
Y Trottier, G Cancel, I An-Gourfinkel, Y Lutz, C Weber, A Brice, E Hirsch, J L Mandel
ABSTRACT
Spinocerebellar ataxia type 3 or Machado-Joseph disease (SCA3/MJD) is an autosomal dominant neurodegenerative disorder caused by an unstable and expanded CAG trinucleotide repeat that leads to the expansion of a polyglutamine tract in a protein of unknown function, ataxin-3. We have generated and characterized a panel of monoclonal and polyclonal antibodies raised against ataxin-3 and used them to analyze its expression and localization. In Hela cells, multiple isoforms are expressed besides the major 55-kDa form. While the majority of ataxin-3 is cytosolic, both immunocytofluorescence and subcellular fractionation studies indicate the presence of ataxin-3, in particular, of some of the minor isoforms, in the nuclear and mitochodrial compartments. We also show that ataxin-3 can be phosphorylated. In the brain, only one ataxin-3 isoform containing the polyglutamine stretch was detected, and normal and mutated proteins were found equally expressed in all patient brain regions analyzed. In most neurons, ataxin-3 had a cytoplasmic, dendritic, and axonal localization. Some neurons presented an additional nuclear localization. Ataxin-3 is widely expressed throughout the brain, with a variable intensity specific for subpopulations of neurons. Its expression is, however, not restricted to regions that show intranuclear inclusions and neurodegeneration in SCA3/MJD.