MilliporeSigma
  • A phase I first-in-human study with tefinostat - a monocyte/macrophage targeted histone deacetylase inhibitor - in patients with advanced haematological malignancies.
A phase I first-in-human study with tefinostat - a monocyte/macrophage targeted histone deacetylase inhibitor - in patients with advanced haematological malignancies.
British journal of haematology (2013-05-08)
Gert J Ossenkoppele, Bob Lowenberg, Pierre Zachee, Norbert Vey, Dimitri Breems, Arjan A Van de Loosdrecht, Alan H Davidson, Graham Wells, Lindsey Needham, Lindsay Bawden, Martin Toal, Leon Hooftman, Phillip M Debnam
ABSTRACT
Tefinostat (CHR-2845) is a monocyte/macrophage targeted histone deacetylase inhibitor (HDACi). This first-in-human, standard 3 + 3 dose escalating trial of oral, once daily tefinostat was conducted to determine the safety, tolerability, pharmacokinetic and pharmacodynamic profile of tefinostat in relapsed/refractory haematological diseases. Eighteen patients were enrolled at doses of 20-640 mg. Plasma concentrations of tefinostat exceeded those demonstrated to give in vitro anti-proliferative activity. Flow cytometric pharmacodynamic assays demonstrated monocyte-targeted increases in protein acetylation, without corresponding changes in lymphocytes. Dose-limiting toxicities (DLTs) were not observed and dose escalation was halted at 640 mg without identification of the maximum tolerated dose. Drug-related toxicities were largely Common Toxicity Criteria for Adverse Events grade 1/2 and included nausea, anorexia, fatigue, constipation, rash and increased blood creatinine. A patient with chronic monomyelocytic leukaemia achieved a bone marrow response, with no change in peripheral monocytes. An acute myeloid leukaemia type M2 patient showed a >50% decrease in bone marrow blasts and clearance of peripheral blasts. In conclusion, tefinostat produces monocyte-targeted HDACi activity and is well tolerated, without the DLTs, e.g. fatigue, diarrhoea, thrombocytopenia, commonly seen with non-targeted HDACi. The early signs of efficacy and absence of significant toxicity warrant further evaluation of tefinostat in larger studies. (clinicaltrials.gov identifier: NCT00820508).
Sigma-Aldrich
IgG from human serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder
Sigma-Aldrich
Anti-Mouse IgG (Fab specific) F(ab′)2 fragment–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution