MilliporeSigma
  • Aerogels made of chitosan and chondroitin sulfate at high degree of neutralization: Biological properties toward wound healing.
Aerogels made of chitosan and chondroitin sulfate at high degree of neutralization: Biological properties toward wound healing.
Journal of biomedical materials research. Part B, Applied biomaterials (2018-02-10)
Miguel Concha, Alejandra Vidal, Annesi Giacaman, Javier Ojeda, Francisca Pavicic, Felipe A Oyarzun-Ampuero, César Torres, Marcela Cabrera, Ignacio Moreno-Villoslada, Sandra L Orellana
ABSTRACT
In this study, highly neutralized, highly porous, and ultralight polymeric aerogels prepared from aqueous colloidal suspensions of chitosan (CS) and chondroitin sulfate (ChS) nanocomplexes, formulated as quasi-equimolar amounts of both, are described. These aerogels were designed as healing agents under the inspiration of minimizing the amount of matter applied to wounds, reducing the electrostatic potential of the material and avoiding covalent cross-linkers in order to decrease metabolic stress over wounds. Aerogels synthesized under these criteria are biocompatible and provide specific properties for the induction of wound healing. They do not affect neither the metabolic activity of cultured 3T3 fibroblasts nor the biochemical parameters of experimental animals, open wounds close significantly faster and, unlike control wounds, complete reepithelialization and scarring can be attained 14 days after surgery. Because of its hydration abilities, rapid adaptation to the wound bed and the early accelerator effect of wound closure, the CS/ChS aerogels appear to be functional inducers of the healing. Previous information show that CS/ChS aerogels improve wound bed quality, increase granulation tissue and have pain suppressive effect. CS/ChS aerogels are useful as safe, inexpensive and easy to handle materials for topical applications, such as skin chronic wounds. © 2018 Wiley Periodicals, Inc. J Biomed Mater Res Part B: Appl Biomater, 106B: 2464-2471, 2018.
Sigma-Aldrich
(±)-2-(p-Methoxyphenoxy)propionic acid, ≥98%