MilliporeSigma
  • Characterization of a Long Non-Coding RNA, the Antisense RNA of Na/K-ATPase α1 in Human Kidney Cells.
Characterization of a Long Non-Coding RNA, the Antisense RNA of Na/K-ATPase α1 in Human Kidney Cells.
International journal of molecular sciences (2018-07-25)
Xiaoming Fan, Usman M Ashraf, Christopher A Drummond, Huilin Shi, Xiaolu Zhang, Sivarajan Kumarasamy, Jiang Tian
ABSTRACT
Non-coding RNAs are important regulators of protein-coding genes. The current study characterized an antisense long non-coding RNA, ATP1A1-AS1, which is located on the opposite strand of the Na/K-ATPase α1 gene. Our results show that four splice variants are expressed in human adult kidney cells (HK2 cells) and embryonic kidney cells (HEK293 cells). These variants can be detected in both cytosol and nuclear fractions. We also found that the inhibition of DNA methylation has a differential effect on the expression of ATP1A1-AS1 and its sense gene. To investigate the physiological role of this antisense gene, we overexpressed the ATP1A1-AS1 transcripts, and examined their effect on Na/K-ATPase expression and related signaling function in human kidney cells. The results showed that overexpression of the ATP1A1-AS1-203 transcript in HK2 cells reduced the Na/K-ATPase α1 (ATP1A1) gene expression by approximately 20% (p < 0.05), while reducing the Na/K-ATPase α1 protein synthesis by approximately 22% (p < 0.05). Importantly, overexpression of the antisense RNA transcript attenuated ouabain-induced Src activation in HK2 cells. It also inhibited the cell proliferation and potentiated ouabain-induced cell death. These results demonstrate that the ATP1A1-AS1 gene is a moderate negative regulator of Na/K-ATPase α1, and can modulate Na/K-ATPase-related signaling pathways in human kidney cells.
Sigma-Aldrich
Anti-phospho-Src (pTyr418) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution