MilliporeSigma
  • A Small β-Carboline Derivative "B-9-3" Modulates TGF-β Signaling Pathway Causing Tumor Regression in Vivo.
A Small β-Carboline Derivative "B-9-3" Modulates TGF-β Signaling Pathway Causing Tumor Regression in Vivo.
Frontiers in pharmacology (2018-08-07)
Hui Zhong, Abdelkader Daoud, Jichun Han, Xiaohong An, Caili Qiao, Lanlan Duan, Yichuan Wang, Zhenfeng Chen, Jia Zhou, Jing Shang
ABSTRACT
Targeting tumor microenvironment (TME) is crucial in order to overcome the anti-cancer therapy resistance. In this study, we report the antitumor activity of a newly synthesized β-carboline derivative "B-9-3." Here, this small molecule showed a promising antitumor activity in vivo along with an enhanced immune response as reflected by a reduction of regulatory T cells and increased CD4+/CD8+ T cells. Further, B-9-3 decreased the number of myofibroblasts not only in the tumor but also in the lung suggesting an anti-metastatic action. The reduction of myofibroblasts was associated with lower expression of epithelial-to-mesenchymal transition markers and a decrease of phosphorylated SMAD2/3 complex indicating the implication of TGF-β signaling pathway in B-9-3's effect. The blockade of myofibroblasts induction by B-9-3 was also verified in vitro in human fibroblasts treated with TGF-β. To elucidate the mechanism of B-9-3's action on TGF-β pathway, first, we investigated the molecular interaction between B-9-3 and TGF-β receptors using docking method. Data showed a weak interaction of B-9-3 with the ATP-binding pocket of TGFβRI but a strong one with a ternary complex formed of extracellular domains of TGFβRI, TGFβRII, and TGF-β. In addition, the role of TGFβRI and TGFβRII in B-9-3's activity was explored in vitro. B-9-3 did not decrease any of the two receptors' protein level and only reduced phosphorylated SMAD2/3 suggesting that its effect was more probably due to its interaction with the ternary complex rather than decreasing the expression of TGF-β receptors or interfering with their ATP-binding domains. B-9-3 is a small active molecule which acts on the TGF-β signaling pathway and improves the TME to inhibit the proliferation and the metastasis of the tumor with the potential for clinical application.