MilliporeSigma
  • Low Protease Content in Medicago truncatula Cell Cultures Facilitates Recombinant Protein Production.
Low Protease Content in Medicago truncatula Cell Cultures Facilitates Recombinant Protein Production.
Biotechnology journal (2018-03-13)
Rita B Santos, Balakumaran Chandrasekar, Manoj K Mandal, Farnusch Kaschani, Markus Kaiser, Leonard Both, Renier A L van der Hoorn, Andreas Schiermeyer, Rita Abranches
ABSTRACT
Medicago truncatula is an established model for studying legume biology. More recently, it has also been exploited as a Molecular Farming platform for the production of recombinant proteins, with the successful expression of fungal and human proteins in plants and cell suspension cultures of this species. One of the challenges that now must be overcome is the degradation of final products during production and downstream processing stages. In the M. truncatula genome, there are more than 400 putative protease-encoding genes, but to date, the proteolytic content of Medicago cell cultures has not been studied. In this report, the proteolytic activities that can potentially hamper the successful production of recombinant proteins in this system are evaluated. The potential proteases responsible for the degradation of target proteins are identified. Interestingly, the number of proteases found in Medicago spent medium is considerably lower than that of the well-established tobacco bright yellow 2 (BY-2) system. Papain-like cysteine proteases are found to be the major contributors to recombinant protein degradation in Medicago. This knowledge is used to engineer a cell line with reduced endogenous protease activity by expressing a selective protease inhibitor, further improving this expression platform.
Sigma-Aldrich
ExtrAvidin®−Cy3, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution