MilliporeSigma
  • miR-129 inhibits tumor growth and potentiates chemosensitivity of neuroblastoma by targeting MYO10.
miR-129 inhibits tumor growth and potentiates chemosensitivity of neuroblastoma by targeting MYO10.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2018-06-06)
Xiqian Wang, Jing Li, Xiao Xu, Jiachun Zheng, Qingbo Li
ABSTRACT
Although the treatment strategies for neuroblastoma (NB) develop rapidly, a considerable number of patients could not benefit from chemotherapy. Here, we revealed a miR-129-MYO10 axis that regulated neuroblastoma growth and chemosensitivity. Mechanistically, MYO10 was up-regulated in neuroblastoma tissues and associated with poor overall survival. While overexpression of MYO10 enhanced tumor growth, genetic inhibition of MYO10 led to growth-inhibitory and chemopotentiating effects in neuroblastoma. MYO10 was further identified as a target of miR-129. Our data showed that miR-129 down-regulated MYO10 expression and subsequently suppressed cell growth. Re-expression of MYO10 significantly rescued miR129-mediated proliferation repression and chemosensitivity. In conclusion, our results demonstrated that miR-129 inhibited neuroblastoma growth and potentiated chemosensitivity by targeting MYO10, which may represent promising targets and rational therapeutic options for neuroblastoma.