MilliporeSigma
  • Hypothermia modulates the DNA damage response to ionizing radiation in human peripheral blood lymphocytes.
Hypothermia modulates the DNA damage response to ionizing radiation in human peripheral blood lymphocytes.
International journal of radiation biology (2018-04-19)
Halina Lisowska, Lei Cheng, Alice Sollazzo, Lovisa Lundholm, Aneta Wegierek-Ciuk, Sylwester Sommer, Anna Lankoff, Andrzej Wojcik
ABSTRACT
Low temperature at exposure has been shown to act in a radioprotective manner at the level of cytogenetic damage. It was suggested to be due to an effective transformation of DNA damage to chromosomal damage at low temperature. The purpose of the study was to analyze the kinetics of aberration formation during the first hours after exposing human peripheral blood lymphocytes to ionizing radiation at 0.8 °C and 37 °C. To this end, we applied the technique of premature chromosome condensation. In addition, DNA damage response was analyzed by measuring the levels of phosphorylated DNA damage responsive proteins ATM, DNA-PK and p53 and mRNA levels of the radiation-responsive genes BBC3, FDXR, GADD45A, XPC, MDM2 and CDKN1A. A consistently lower frequency of chromosomal breaks was observed in cells exposed at 0.8 °C as compared to 37 °C already after 30 minutes postexposure. This effect was accompanied by elevated levels of phosphorylated ATM and DNA-PK proteins and a reduced immediate level of phosphorylated p53 and of the responsive genes. Low temperature at exposure appears to promote DNA repair leading to reduced transformation of DNA damage to chromosomal aberrations.
Sigma-Aldrich
6-Phosphonohexanoic acid, 97%
Sigma-Aldrich
Pyridinium chlorochromate, 98%