MilliporeSigma
  • Kinesin 6 Regulation in Drosophila Female Meiosis by the Non-conserved N- and C- Terminal Domains.
Kinesin 6 Regulation in Drosophila Female Meiosis by the Non-conserved N- and C- Terminal Domains.
G3 (Bethesda, Md.) (2018-03-09)
Arunika Das, Jeffry Cesario, Anna Maria Hinman, Janet K Jang, Kim S McKim
ABSTRACT
Bipolar spindle assembly occurs in the absence of centrosomes in the oocytes of most organisms. In the absence of centrosomes in Drosophila oocytes, we have proposed that the kinesin 6 Subito, a MKLP-2 homolog, is required for establishing spindle bipolarity and chromosome biorientation by assembling a robust central spindle during prometaphase I. Although the functions of the conserved motor domains of kinesins is well studied, less is known about the contribution of the poorly conserved N- and C- terminal domains to motor function. In this study, we have investigated the contribution of these domains to kinesin 6 functions in meiosis and early embryonic development. We found that the N-terminal domain has antagonistic elements that regulate localization of the motor to microtubules. Other parts of the N- and C-terminal domains are not required for microtubule localization but are required for motor function. Some of these elements of Subito are more important for either mitosis or meiosis, as revealed by separation-of-function mutants. One of the functions for both the N- and C-terminals domains is to restrict the CPC to the central spindle in a ring around the chromosomes. We also provide evidence that CDK1 phosphorylation of Subito regulates its activity associated with homolog bi-orientation. These results suggest the N- and C-terminal domains of Subito, while not required for localization to the central spindle microtubules, have important roles regulating Subito, by interacting with other spindle proteins and promoting activities such as bipolar spindle formation and homologous chromosome bi-orientation during meiosis.
Sigma-Aldrich
Anti-α-Tubulin−FITC antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture