MilliporeSigma
  • Micropropagation of photinia employing rhizobacteria to promote root development.
Micropropagation of photinia employing rhizobacteria to promote root development.
Plant cell reports (2007-01-06)
Ezequiel E Larraburu, Susana M Carletti, Enrique A Rodríguez Cáceres, Berta E Llorente
ABSTRACT
An alternative protocol was developed for in vitro propagation of photinia (Photinia x fraseri Dress), an ornamental shrub, using the plant growth-promoting rhizobacteria (PGPR) Azospirillum brasilense and Azotobacter chroococcum during rhizogenesis. Shoot tips from four-year-old mature plants, cut in spring and summer, were used as initial explants. They were cultured on Murashige-Skoog (MS) medium with Gamborg's vitamins, N(6)-benzyladenine (BA: 11.1 microM) and gibberellic acid (GA(3): 1.3 microM), obtaining 63% of established explants. The highest shoot length (22.9 mm) and multiplication rate (4.3) was achieved by cultivating for four weeks in the same basal medium supplemented with 4.4 microM BA. Both auxin induction and bacterial inoculation were used for rooting. Elongated shoots were treated with two concentrations of indole-3-butyric acid (IBA: 4.9 or 49.2 microM) during 6 days for auxin induction. Then, the shoots were transferred to an auxin-free medium and inoculated with A. brasilense Cd, Sp7 or A. chroococcum (local strain). Bacterial inoculation induced earlier rooting of photinia shoots. A. brasilense Cd with 49.2 microM IBA pulse showed a significant increase (P <or= 0.05) in root fresh and dry weight (105%, 137%), root surface area (65%) and shoot fresh and dry weight (32%, 62%). A. brasilense Sp7 enhanced the root fresh weight (34%) and root surface area (41%) while no significant differences with A. chroococcum inoculation were detected. The PGPR inoculated micro-cuttings in combination with auxin induction pulses may play a useful role in root organogenesis of micropropagated plants.
Sigma-Aldrich
Gibberellin A4, suitable for plant cell culture, BioReagent, ≥90%
Sigma-Aldrich
Gibberellic acid, suitable for plant cell culture, BioReagent, ≥90% gibberellin A3 basis (of total gibberellins.)
Sigma-Aldrich
Gibberellic acid potassium salt, suitable for plant cell culture, BioReagent, ~95%, ≥50% total GA3 basis