MilliporeSigma
  • TP53INP2 regulates adiposity by activating β-catenin through autophagy-dependent sequestration of GSK3β.
TP53INP2 regulates adiposity by activating β-catenin through autophagy-dependent sequestration of GSK3β.
Nature cell biology (2018-03-30)
Montserrat Romero, Alba Sabaté-Pérez, Víctor A Francis, Ignacio Castrillón-Rodriguez, Ángels Díaz-Ramos, Manuela Sánchez-Feutrie, Xavier Durán, Manuel Palacín, José María Moreno-Navarrete, Birgit Gustafson, Ann Hammarstedt, José Manuel Fernández-Real, Joan Vendrell, Ulf Smith, Antonio Zorzano
ABSTRACT
Excessive fat accumulation is a major risk factor for the development of type 2 diabetes mellitus and other common conditions, including cardiovascular disease and certain types of cancer. Here, we identify a mechanism that regulates adiposity based on the activator of autophagy TP53INP2. We report that TP53INP2 is a negative regulator of adipogenesis in human and mouse preadipocytes. In keeping with this, TP53INP2 ablation in mice caused enhanced adiposity, which was characterized by greater cellularity of subcutaneous adipose tissue and increased expression of master adipogenic genes. TP53INP2 modulates adipogenesis through autophagy-dependent sequestration of GSK3β into late endosomes. GSK3β sequestration was also dependent on ESCRT activity. As a result, TP53INP2 promotes greater β-catenin levels and induces the transcriptional activity of TCF/LEF transcription factors. These results demonstrate a link between autophagy, sequestration of GSK3β into late endosomes and inhibition of adipogenesis in vivo.
Sigma-Aldrich
Fluoromount Aqueous Mounting Medium, for use with fluorescent dye-stained tissues
Sigma-Aldrich
Pioglitazone
Sigma-Aldrich
Complete Whole Transcriptome Amplification Kit, DNA polymerase included, Complete Kit with optimized enzyme to amplify total RNA in <4 hours, no 3′ bias