MilliporeSigma
  • Somatic Mutational Landscape of Splicing Factor Genes and Their Functional Consequences across 33 Cancer Types.
Somatic Mutational Landscape of Splicing Factor Genes and Their Functional Consequences across 33 Cancer Types.
Cell reports (2018-04-05)
Michael Seiler, Shouyong Peng, Anant A Agrawal, James Palacino, Teng Teng, Ping Zhu, Peter G Smith, Silvia Buonamici, Lihua Yu
ABSTRACT
Hotspot mutations in splicing factor genes have been recently reported at high frequency in hematological malignancies, suggesting the importance of RNA splicing in cancer. We analyzed whole-exome sequencing data across 33 tumor types in The Cancer Genome Atlas (TCGA), and we identified 119 splicing factor genes with significant non-silent mutation patterns, including mutation over-representation, recurrent loss of function (tumor suppressor-like), or hotspot mutation profile (oncogene-like). Furthermore, RNA sequencing analysis revealed altered splicing events associated with selected splicing factor mutations. In addition, we were able to identify common gene pathway profiles associated with the presence of these mutations. Our analysis suggests that somatic alteration of genes involved in the RNA-splicing process is common in cancer and may represent an underappreciated hallmark of tumorigenesis.
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human FUBP1