Skip to Content
MilliporeSigma
Undecyl alcohol 97%, FG

Undecyl alcohol

Undecyl alcohol 97%, FG
Synonym(s):
1-Undecanol, Alcohol C11, Undecyl alcohol
Linear Formula:
CH3(CH2)10OH
CAS Number:
Molecular Weight:
172.31
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing