Tryptic Soy Agar - Ready-to-use Strips
Products
Product Category
Brand
Physical Form
Product Line
Chemical Composition
Compatibility
Quality Segment
Application
Microorganism Suitability
Package Size
Sterility
Sterilization Method
Showing 1-3 of 3
Compare
Product Number
Description
Pricing