Skip to Content
MilliporeSigma
TRIS OmniPur® Grade, >=99.8%

TRIS

TRIS OmniPur® Grade, >=99.8%
Synonym(s):
Tris(hydroxymethyl)aminomethane, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Trometamol
Linear Formula:
NH2C(CH2OH)3
CAS Number:
Molecular Weight:
121.14

Products

Product Category
Brand
Boiling Point (°C)
Color
Formula Weight
Melting Point (°C)
pH Value
Physical Form
pK Value
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing