Skip to Content
MilliporeSigma
trans-Zeatin-riboside suitable for plant cell culture, BioReagent, ~95%

trans-Zeatin-riboside

trans-Zeatin-riboside suitable for plant cell culture, BioReagent, ~95%
Synonym(s):
9-(β-D-Ribofuranosyl)-trans-zeatin, N6-(trans-4-Hydroxy-3-methyl-2-buten-1-yl)adenosine
Empirical Formula (Hill Notation):
C15H21N5O5
CAS Number:
Molecular Weight:
351.36

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Product Line
Purity
Application
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing