Skip to Content
MilliporeSigma
Acetaldehyde solution natural, 50 wt. % in ethanol, analytical standard

Acetaldehyde

Acetaldehyde solution natural, 50 wt. % in ethanol, analytical standard
Synonym(s):
Ethanal, Acetaldehyde
Linear Formula:
CH3CHO
CAS Number:
Molecular Weight:
44.05

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique