Skip to Content
MilliporeSigma
Biotin for system suitability European Pharmacopoeia (EP) Reference Standard

Biotin

Biotin for system suitability European Pharmacopoeia (EP) Reference Standard
Synonym(s):
D-Biotin, Bios II, Coenzyme R, Vitamin B7, Vitamin H, Biotin
Empirical Formula (Hill Notation):
C10H16N2O3S
CAS Number:
Molecular Weight:
244.31
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique