Skip to Content
MilliporeSigma
L-Isoleucine certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

L-Isoleucine

L-Isoleucine certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
(2S,3S)-2-Amino-3-methylpentanoic acid, L-Isoleucine
Linear Formula:
C2H5CH(CH3)CH(NH2)CO2H
CAS Number:
Molecular Weight:
131.17
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Agency
Format
Analyte Suitability
Quality Segment
Application
Optical Isomer
API Family
Reference Material Type
Special Grade
Technique