Valacyclovir Related Compound G
Synonym(s):
4-(Dimethylamino)pyridine, N,N-Dimethylpyridin-4-amine, DMAP
Empirical Formula (Hill Notation):
C7H10N2
CAS Number:
Molecular Weight:
122.17
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing