Skip to Content
MilliporeSigma
Trypan Blue solution 0.4%, for microscopy

Trypan Blue solution

Trypan Blue solution 0.4%, for microscopy
Empirical Formula (Hill Notation):
C34H24N6Na4O14S4
CAS Number:
Molecular Weight:
960.81

Products

Brand
Color
Formula Weight
Physical Form
Quality Segment
Application
Sterilization Method
Technique
Showing 1-2 of 2