Skip to Content
MilliporeSigma
Sputolysin® Reagent Intended for the isolation of epithelial cells, pathogenic or saprophytic bacteria, fungi, and yeasts from sputum.

DL-Dithiothreitol solution

Sputolysin® Reagent Intended for the isolation of epithelial cells, pathogenic or saprophytic bacteria, fungi, and yeasts from sputum.
Synonym(s):
(±)-threo-1,4-Dimercapto-2,3-butanediol solution, DTT
Empirical Formula (Hill Notation):
C4H10O2S2
CAS Number:
Molecular Weight:
154.25

Products

Brand
Formula Weight
pH Value
Physical Form
Product Line
Research Area
Quality Segment
Reagent Type