Skip to Content
MilliporeSigma
Polyoxyethylene (20) sorbitan monolaurate solution ampule, ~10% in H2O

TWEEN® 20

Polyoxyethylene (20) sorbitan monolaurate solution ampule, ~10% in H2O
Synonym(s):
TWEEN 20, TWEEN® 20, Polyethylene glycol sorbitan monolaurate, Polyoxyethylenesorbitan monolaurate, Polyethylene glycol sorbitan monolaurate solution
CAS Number:

Products

Product Category
Brand
Manufacturer
Physical Form
Quality Segment
Application
Technique