![Propyl parahydroxybenzoate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/234/384/3b17a307-f459-49d4-a9f2-19569e622ff6/640/3b17a307-f459-49d4-a9f2-19569e622ff6.png)
Propyl parahydroxybenzoate
![Propyl parahydroxybenzoate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/234/384/3b17a307-f459-49d4-a9f2-19569e622ff6/640/3b17a307-f459-49d4-a9f2-19569e622ff6.png)
Synonym(s):
Propyl 4-hydroxybenzoate, 4-Hydroxybenzoic acid propyl ester, Propyl paraben
Linear Formula:
HOC6H4CO2CH2CH2CH3
CAS Number:
Molecular Weight:
180.20
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing