Skip to Content
MilliporeSigma
N4,N4,N4′,N4′-Tetra[(1,1′-biphenyl)-4-yl]-(1,1′-biphenyl)-4,4′-diamine ≥99% (HPLC)

N4,N4,N4′,N4′-Tetra[(1,1′-biphenyl)-4-yl]-(1,1′-biphenyl)-4,4′-diamine

N4,N4,N4′,N4′-Tetra[(1,1′-biphenyl)-4-yl]-(1,1′-biphenyl)-4,4′-diamine ≥99% (HPLC)
Synonym(s):
N,N′-Tetra(4-biphenyl)benzidine
Empirical Formula (Hill Notation):
C60H44N2
CAS Number:
Molecular Weight:
793.00

Products

Brand
Formula Weight
Quality Segment
Showing 1-1 of 1