Skip to Content
MilliporeSigma
Lupeol analytical standard

Lupeol

Lupeol analytical standard
Synonym(s):
β-Viscol, 20(29)-Lupen-3β-ol, 3β-Hydroxy-20(29)-lupene, Clerodol, Fagarasterol, Lupenol, Monogynol B
Empirical Formula (Hill Notation):
C30H50O
CAS Number:
Molecular Weight:
426.72
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing