Endfitting with fixed 1 μm frit for TSKgel® Super Series RP and SuperH2 Columns
Synonym(s):
TSKgel® Hardware and Accessories
Products
Manufacturer
Product Line
Package Size
Pore size (µm)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing