![Dimethyl 2-[(2-aminoethoxy)methyl]-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/632/583/0b21b85f-09ba-46d1-9cc7-482d7a705354/640/0b21b85f-09ba-46d1-9cc7-482d7a705354.png)
Dimethyl 2-[(2-aminoethoxy)methyl]-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate
![Dimethyl 2-[(2-aminoethoxy)methyl]-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/632/583/0b21b85f-09ba-46d1-9cc7-482d7a705354/640/0b21b85f-09ba-46d1-9cc7-482d7a705354.png)
Empirical Formula (Hill Notation):
C19H23ClN2O5
CAS Number:
Molecular Weight:
394.85
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Special Grade
Showing 1-1 of 1