Corning® Transwell® polycarbonate membrane cell culture inserts
Products
Product Category
Manufacturer
Binding Type
Diameter (mm)
Surface Area cm²
Wells
Working Volume (ml)
Color
Membrane Material
Package Size
Pore size (µm)
Sterility
Technique
Showing 1-11 of 11
Compare
Product Number
Description
Pricing