Skip to Content
MilliporeSigma
L-Alanine ≥98% (TLC)

L-Alanine

L-Alanine ≥98% (TLC)
Synonym(s):
(S)-2-Aminopropionic acid, L-α-Aminopropionic acid, L-Alanine, (S)-(+)-Alanine
Empirical Formula (Hill Notation):
C3H7NO2
CAS Number:
Molecular Weight:
89.09

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Gene Alias
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Optical Isomer
Amino Acid
Technique