Skip to Content
MilliporeSigma
Imidazole for molecular biology, ≥99% (titration), free-flowing, Redi-Dri™

Imidazole

Imidazole for molecular biology, ≥99% (titration), free-flowing, Redi-Dri™
Synonym(s):
1,3-Diaza-2,4-cyclopentadiene, Glyoxaline, Imidazole, OmniPur® Imidazole, Imidazole, ULTROL® Grade
Linear Formula:
-NHCH=NCH=CH-
CAS Number:
Molecular Weight:
68.08

Products

Product Category
Brand
Boiling Point (°C)
Color
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
pK Value
Product Line
Purity
Research Area
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technique